Valve Replacement Parts for Shimadzu LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8001-0603 8001-0614 8001-0809
CAAG8001-0601EA 324.86 CAD
CAAG8001-0601 CAAG8001-0603 CAAG8001-0608 CAAG8001-0614 CAAG8001-0809
Valve Replacement Parts for Shimadzu LC Systems, Agilent Technologies
High-quality replacement valve parts for selected Shimadzu HPLC and LC/MS instruments.
  • PEEK offers high resistance, versatility, and applicability for the entire pH range between 0 and 14
  • Stators typically require replacement only if the ports or sealing surfaces become damaged, which can be prevented by using proper injection needles
Agilent valve supplies work seamlessly with selected Shimadzu HPLC systems for reliable, accurate, and reproducible results. Stators are available in 316 stainless steel, PEEK, and ceramic. Rotor seals are available in PEEK for a wide range of applications.