Spray Chambers and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0329 8003-0334 8003-0462 8003-0817
CAAG8003-0329EA 1948.89 CAD
CAAG8003-0329 CAAG8003-0334 CAAG8003-0462 CAAG8003-0817
Spray Chambers and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Full portfolio of ICP-OES spray chambers compatible with Perkin Elmer ICP-OES systems.
  • Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent spray chambers and supplies are fully compatible with PerkinElmer Optima ICP-OES. The single pass glass cyclonic spray chamber for good signal-to-noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
The new Agilent inert spray chamber is compatible with the Avio 200 and 500 ICP-OES. It features a durable sandblasted coating and removable internal baffle. The sanblasted coating removes the need for chemical coatings which need periodic rejuvination. The removable internal baffle allows the spray chamber to be used as a single pass or double pass spray chamber essential being 2 spray chambers in one.