Special Function Valves for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5043-0269 5067-4244 5067-4266 5067-5424 5067-5425 5067-5426 5067-5437 5068-0115 5068-0239 5068-0240 5068-0257
CAAG5043-0269EA 46.88 CAD
CAAG5043-0269 CAAG5067-4244 CAAG5067-4266 CAAG5067-5424 CAAG5067-5425 CAAG5067-5426 CAAG5067-5437 CAAG5068-0115 CAAG5068-0239 CAAG5068-0240 CAAG5068-0257
Special Function Valves for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
High-quality supplies for special function valves such as 2D-LC valves.
  • Supplies for 2D-LC valve heads;2D-LC active flow modulation valves and supplies
  • Stream selector valves and supplies
The Agilent InfinityLab 2D-LC solution brings the separation power of two-dimensional LC (2D-LC) to every lab. This solution makes it easy to switch between one-dimensional UHPLC and all modes of 2D-LC, allowing you to adjust separation performance to your sample complexity, as well as guaranteeing highest usage of a single system.
Easy-to-use software enables fast method setup for heart-cutting 2D-LC, comprehensive 2D-LC, multiple heart-cutting 2D-LC, or Agilent's unique high-resolution sampling 2D-LC.
High-quality supplies, including rotor seals, stator heads and bearing rings, fit perfectly to your 2D-LC valves, ensuring optimal performance and minimal system downtime.