Smart SPME Arrows, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5610-5858 5610-5859 5610-5860 5610-5861 5610-5862 5610-5863 5610-5864 5610-5865 5610-5866 5610-5867 5610-5869
CAAG5610-5858PK 3860.25 CAD
CAAG5610-5858 CAAG5610-5859 CAAG5610-5860 CAAG5610-5861 CAAG5610-5862 CAAG5610-5863 CAAG5610-5864 CAAG5610-5865 CAAG5610-5866 CAAG5610-5867 CAAG5610-5869
Smart SPME Arrows, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Smart SPME Arrows for PAL3 instrumentation.
  • Up to 10 times more sensitive than SPME fibers
  • Smart technology enables critical parameter tracking for each unique SPME Arrow
  • Automatic application of the correct parameters for the individual smart SPME Arrow
  • Compatible with all PAL3 models and existing SPME tools
  • Color coded to indicate the Arrow coating and film thickness
  • Preinstalled with holder for easy maintenance
  • Smart SPME Arrow kits available for method development
Agilent offers smart SPME Arrows for compatibility with all PAL3 models including PAL 3 Series II smart instrumentation. SPME Arrows can be used in headspace analysis or by liquid immersion to concentrate an analyte onto the smart SPME Arrow. Each smart SPME Arrow is equipped with a unique smart chip containing parameters, ranges, and usage history for the specific Arrow. Agilent smart SPME Arrows are color coded to easily identify chemistry and thickness. Whether you are a new SPME Arrow user or switching to smart technology, you can be confident that performance of Agilent smart SPME Arrows remains of the highest quality.