Sample Compartment Windows for Cary FTIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
210228390 PIKE-160-1009 PIKE-160-1010 PIKE-160-1015 PIKE-160-1125 PIKE-160-1126 PIKE-160-1127 PIKE-160-1133
CAAG210228390EA 1919.85 CAD
CAAG210228390 CAGPIKE1601009 CAGPIKE1601010 CAGPIKE1601015 CAGPIKE1601125 CAGPIKE1601126 CAGPIKE1601127 CAGPIKE1601133
Sample Compartment Windows for Cary FTIR Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent offers a wide range of high quality window materials to suit your analytical needs, such as KBr, KRS5 and ZnSe.
  • Ideal for FTIR Spectroscopy
  • Excellent transmission (wavelength range depends on material type)​
  • Good resistance to mechanical shock;Windows are available in standard sizes
  • Windows are individually packaged and silica gel is included to protect windows from exposure to humidity
Important considerations when selecting a window are the wavelength range transmitted, solubility, reactivity, the refractive index, and the mechanical and thermal characteristics.