QuickSwap Restrictors for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3185-60300 G3185-60362
CAAGG318560300EA 451.09 CAD
CAAGG318560300 CAAGG318560362
QuickSwap Restrictors for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
QuickSwap interface restrictors can increase the productivity of your 5973N and 5975 inert MSD systems, allowing you to change columns without venting the MSD.
  • Prefabricated for convenience and ease of use
  • For applications requiring other restrictor sizes, Agilent offers a variety of deactivated fused silica tubing, SilTitle ferrules and swaging tools
Agilent QuickSwap capillary flow technology module and preswaged, fused silica tubing restrictors can increase the productivity of your Agilent 5973N and 5975 inert MSD systems, allowing you to changeMore Product Information