QuickProbe GC/MS System Consumables, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5190-5104 5190-5118 G3971-20251 G3971-60200
CAAG5190-5104EA 94.22 CAD
CAAG5190-5104 CAAG5190-5118 CAAGG397120251 CAAGG397160200
QuickProbe GC/MS System Consumables, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
QuickProbe GC/MS system consumables include insertion probes, columns, and specialized liners.
  • Unique fritted liner, 4 mm int.Ø
  • Two probe holders and three probe designs for unique sample types
  • Insertion probe round tip (recommended for liquids, dissolved samples, and tablets), 100/pk, used with QuickProbe probe holder
  • Insertion probe pocket tip (recommended for powders), 100/pk, used with QuickProbe probe holder
  • TSP solid probe vial (recommended for solid samples), 100/pk, used with TSP probe holder
  • QuickProbe DB-1HT, 0.25 mm × 0.1 μm × 2 m, for use in
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable