Purge Extension Windows for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0384 8003-0386 8003-0387 8003-0394 8003-0472
CAAG8003-0384EA 438.25 CAD
CAAG8003-0384 CAAG8003-0386 CAAG8003-0387 CAAG8003-0394 CAAG8003-0472
Purge Extension Windows for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Axial and Radial purge extension windows for Perkin Elmer Optima ICP-OES.
  • Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and
More Product Information