Nebulizers and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0320 8003-0326 8003-0327 8003-0974 G3139-68000
CAAG8003-0320EA 1856.8 CAD
CAAG8003-0320 CAAG8003-0326 CAAG8003-0327 CAAG8003-0974 CAAGG313968000
Nebulizers and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Full portfolio of ICP-OES nebulizers provide optimal performance with your Perkin Elmer instrument.
  • Manufactured to fine tolerances for reproducible and stable sample uptake
  • Tight specifications deliver fine droplets for best signal intensity and optimum signal to noise
  • Fewer repeats means less downtime and improved productivity;Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime;Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide best-in-class technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent has a wide selection of Perkin Elmer supplies guarenteed to fit with your Perkin Elmer ICP-OES including the Agilent designed OneNeb 2 nebulizer. Agilent's nebulizers for PerkinElmer ICP-OES systems enhance performance productivity. Tight specifications for delivering fine droplets improve signal intensity and signal-to-noise. Choose from GemTip, GemCone, Meinhard, or inert MiraMist.
The Agilent OneNeb Series 2 nebulizer improves performance compared to glass concentric and inert types commonly used. The OneNeb Series 2 is up to four times more sensitive than most nebulizers and provides tolerance to high levels of dissolved solids and aggressive acids.