Liquid Chromatography (LC) Interface Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1820-65541 G3280-80602 G3280-80603
CAAGG182065541KT 1330.83 CAD
CAAGG182065541 CAAGG328080602 CAAGG328080603
Liquid Chromatography (LC) Interface Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent is an expert in speciation analysis, offering HPLC-ICP-MS parts and interface supplies.
  • HPLC separation, followed by ICP-MS detection is well established as the speciation analytical method of choice for many sample types
  • HPLC connection kits provide all the components, tubing, connectors, and documentation required to interface an Agilent Infinity LC or other HPLC or IC to the Agilent 7700, 7800, 7900, 8800, 8900 ICP-MS
  • Capillary LC interface components enable rapid sample stabilization and washout times and maintains chromatographic peak
More Product Information