ISIS-DS (Integrated Sample Introduction System - Discrete Sampling) Supplies for ICP-MS (7500), Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3138-65158 G3138-65157
CAAG5064-8015PK 136.55 CAD
CAAG5064-8015 CAAGG313865125 CAAGG313865158 AGG3138-65157
ISIS-DS (Integrated Sample Introduction System - Discrete Sampling) Supplies for ICP-MS (7500), Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies and components for the ISIS-DS for the 7500 ICP-MS.
  • A complete kit of tubing for peristaltic pump plumbing needs
  • A range of standard accessory parts for high sample throughput, discrete sampling are provided
  • Additional consumables may also be purchased such as mixers, sample loops, tubing adapters and plastic trays
Agilent's unique Integrated Sample Introduction System with Discrete Sampling (ISIS-DS) enhances the functionality and flexibility of the Agilent 7500 Series ICP-MS. Designed and manufactured by Agilent, ISIS-DS features high precision peristaltic pumps and an optional a robust switching valve and customizable fixed volume sample loops to allow complete flexibility of configuration.
The ISIS hardware is built into the 7500 chassis and close coupledclose-coupled to increase productivity through high-speed sample uptake and rapid washout. To enhance the capabilities that the system offers and maintain performance even after heavy use, Agilent provides a range of replacement components and other supplies.