Intuvo Tools, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G4581-60260 G4582-20085 G4581-20006 8710-2790 8710-2800 8710-2801 G4580-68300 G4581-20522 G4583-20005 G4586-20027
CAG4581-60260EA 53.03 CAD
CAG4581-60260 CAG4582-20085 CAAGG458120006 CAAG8710-2790 CAAG8710-2800 CAAG8710-2801 CAAGG458068300 CAAGG458120522 CAAGG458320005 CAAGG458620027
Intuvo Tools, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Features and benefits of Intuvo tools.
  • Intuvo torque driver: Ergonomic design provides exact torque for compression bolts
  • For use with the Agilent Intuvo 9000 system
The Intuvo 9000 GC system uses a unique torque driver to secure the flow chip and guard chip. The driver attaches to the door and is included with each instrument. Replacement and back-up tools can be ordered here.
In addition to the torque driver, other tools are available to help operate the Intuvo GC: Tweezers for removing and inserting gaskets. Wrenches for tightening the inlet compression bolt onto the guard chip.
These are also available as part of the Intuvo installation kit (part number 19199U).
* Product only compatible with the Agilent Intuvo 9000 GC system.