Pierce™ Deuterium-labeled Cross Linkers, Thermo Scientific
Supplier: Thermo Scientific
21590 21595 21610
CAPI21590EA 380.13 CAD
CAPI21590 CAPI21595 CAPI21610 CAPI21615
Pierce™ Deuterium-labeled Cross Linkers, Thermo Scientific
Protein Linking and Labeling Systems
Cross linkers are used for the identification of near-neighbor protein relationships and ligand-receptor interactions.
  • Deuterium-labeled cross linkers
Deuterium-labeled cross linkers are specially manufactured for crosslinking protein structure: Function analysis by mass spectrometry.