PROPOR HC MURUS Capsules, Parker Bioscience
Supplier: PARKER HANNIFIN TE
ZLHC1-620BB-PSL ZLHC3-620BB-PNL ZLHC3-620BB-PSL ZLHC3-620BB-PNL ZLHC3-620BB-SL ZLHC1-620BB-PSL ZLHCK-620BB-PNL ZLHC1-620BB-PNL ZLHC2-620BB-PNL ZLHCK-620BB-PSL ZLHC2-620BB-PSL ZLHC2-620TT-PNT ZLHC1-620AA-PNL-S ZLHC1-620AA-PSL-S ZLHC1-620HH-PNL-S ZLHC1-620HH-PSL-S ZLHC2-620AA-PNL-S ZLHC2-620AA-PSL-S ZLHC2-620HH-PNL-S ZLHC2-620HH-PSL-S ZLHC3-620AA-PNL-S ZLHC3-620AA-PSL-S ZLHC3-620HH-PNL-S ZLHC3-620HH-PSL-S ZLHCK-620AA-PNL-S ZLHCK-620AA-PSL-S ZLHCK-620HH-PNL-S ZLHCK-620HH-PSL-S
76541-090EA 1311.08 CAD
76541-090 76541-094 76541-096 76545-582 76545-584 76545-588 76552-166 76552-168 76552-170 76552-172 76552-174 76554-142 77676-198 77676-192 77676-186 77676-208 77676-214 77676-194 77676-204 77676-202 77676-184 77676-190 77676-212 77676-210 77676-196 77676-200 77676-206 77676-188
PROPOR HC MURUS Capsules, Parker Bioscience
Filters Membrane Filter Capsules
PROPOR HC sterilizing grade filters have been specifically designed for the effective and economical processing of difficult to filter solutions.
  • Optimized membrane configuration allows up to 10X the throughput compared to single layer membrane products
  • Integral prefilter layer can condense filter trains for greater processing economy
  • Incorporates a fully validated and integrity testable 0.2 micron membrane for assurance of sterility
  • Low binding for minimal product loss
  • Single-use Capsule format available with Pre-sterile version by Gamma-irradiation