Portable Aspirator, Diaphragm Pump, Mortech
Supplier: Mortech
FH300
76549-894EA 3443.67 CAD
76549-894
Portable Aspirator, Diaphragm Pump, Mortech
Pumps Aspiration systems
Designed to provide suction that is available when you need it. These units are engineered to supply high performance suction capability at a moment’s notice. Model FH300 has a Lumitone® enamel finish with an attractive high impact, fire retardant base.
  • Diaphragm pump unit designed for general suction use
  • High performance, powerful suction capability, when and where you need it at a moment’s notice
  • Lumitone® enamel finish with an attractive, high impact and fire retardant base
  • Accommodates 600 ml glass bottle or 1100 ml disposable canisters
  • Compact for easy storage