Mopec Harmony Tissue Marking Dyes, Mortech
Supplier: Mopec
BG101-250 BG101-60 BG104-250 BG104-60 BG106-250 BG106-60 BG107-250 BG107-60 BG109-250 BG109-60 BG110-250 BG110-60 BG124-250 BG124-60 BG126-250 BG126-60 BG127-250 BG127-60
76549-706EA 226.37 CAD
76549-706 76549-708 76549-710 76549-712 76549-714 76549-716 76549-718 76549-720 76549-722 76549-724 76549-726 76549-728 76549-730 76549-732 76549-734 76549-736 76549-738 76549-740
Mopec Harmony Tissue Marking Dyes, Mortech
Stains Histology Stains
Mopec’s range of Harmony tissue marking dyes are used to permanently mark surgical tissue margins in Histological specimens. Marking dyes are available in nine colors, with a standardized viscosity over the range. This ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color.
  • Emulsified pigments are blended perfectly with an aqueous base to provide a consistent and high-quality product every time
  • Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both prior and after tissue processing
  • Dries in 3 minutes, improving laboratory workflow at the grossing station
  • Nonflammable
  • Available in either 60 ml or 250 ml round neck, induction sealed bottles with a standard flip top lid
Harmony has also been purposely engineered to reduce in viscosity during application, giving users an easy to apply dye without any dripping or running and ensures an even coverage of dye on the specimen.
High concentrations of pigment in the dyes increase their durability and allow diagnostic specimens to be more easily marked. Harmony coats specimens with minimal penetration of the tissue surface, whilst its strong adhesion qualities mean the marking dye will not interfere with diagnostic interpretations, making it ideal for radical prostatectomies.