Dissecting Board, Raised, Mortech
Supplier: Mortech
BC025
76549-602EA 446.89 CAD
76549-602
Dissecting Board, Raised, Mortech
Dissecting Boards
This raised dissection board is made from a high density polyethylene and comes with four 3" legs allowing the board to be raised.
  • Manufactured from high density polyethylene
  • Provides an excellent cutting surface
  • Four 3" legs raise the the dissecting platform