Qlicksmart® Blade Remover, BladeFlask EVO , Mortech
Supplier: Mortech
Qlicksmart®
AJ160
76549-474EA 100.31 CAD
76549-474
Qlicksmart® Blade Remover, BladeFlask EVO , Mortech
Blade Removers and Dispensers
The Qlicksmart® BladeFlask EVO can accommodate even more types of scalpel handles and blades including large autopsy scalpels, thick and round handles, even hexagonal shaped versions – giving you greater freedom in using any scalpel you want. It is so simple to use with just one hand.
  • 100 blade capacity
  • Compatible blades: All commercially available scalpel blades including size PM60A, PM60B, and PM70
  • Compatible handles: All commercially available scalpel handles including PM6 and PM8
  • Easy mounting with optional mounting bracket
  • OSHA compliant
Embrace safety with the new standard in scalpel blade removals.