Ergonomic Scalpel Handles, Premium, Mortech
Supplier: Mopec
AJ108 AJ109 AJ110
76549-398EA 192.29 CAD
76549-398 76549-400 76549-402
Ergonomic Scalpel Handles, Premium, Mortech
Scalpels and Scalpel Assemblies Scalpel Handles
Mopec’s comfortable ergonomic handles will fit all three sizes (#22, #60, and #70) of scalpel blades common to anatomical pathology.
  • #5 Handle fits #6 – #16 blades
  • #6 Handle fits #20 – #26 blades
  • #8 Handle fits #60 – #70 blades
Available in a variety of sizes.