Forceps, Hemostatic, Disposable, Mortech
Supplier: Mopec
AC601 AC602 AC603 AC604
76549-002EA 92.46 CAD
76549-002 76549-004 76549-006 76549-008
Forceps, Hemostatic, Disposable, Mortech
Forceps
Disposable hemostatic forceps ideal for a wide variety of laboratory and post-mortem dissection procedures.
  • Stainless steel
  • Serrated tip
  • Pack of 12
Available with straight or curved edges.
Order Now
SPECIFICATIONS
Environmentally Preferable