Metal Sieves
Supplier: United Scientific Supplies
UNMTSV06
470339-924EA 360.24 CAD
470339-924
Metal Sieves
Sieves General-Purpose Sieves
Stackable steel sieves for separating sediment or particles.
  • Made from durable stainless steel
  • Stackable design
  • Set includes lid and catch pan
These stackable stainless steel mesh sieves are ideal for separating soil or sediment particles of multiple sizes. Available in six convenient sizes with a lid and catch pan. All sieves are 20 cm in diameter and 4cm deep.
Sieve sizes: 4000, 2000, 500, 250, 125, and 63 micron.