VWR® Cleanroom 16" Nitrile Sterile Gloves
Supplier: VWR International
76557-800 76557-802 76557-804 76557-806 76557-808 76557-810
76557-800CS 877.63 CAD
76557-800 76557-802 76557-804 76557-806 76557-808 76557-810
VWR® Cleanroom 16" Nitrile Sterile Gloves
Gloves
VWR® 16" sterile nitrile gloves offer reliable, durable, and comfortable protection for cleanroom applications. Ideal for pharmaceutical, biotech, and electronics industries, these gloves meet the demands of controlled environments by providing excellent barrier protection and comfort during critical tasks.
  • Sterile, designed for elbow length protection
  • Ambidextrous, designed with micro textured fingers and palm, beaded cuff
  • Not made from natural rubber latex
  • Static-dissipative
  • Single-use only, ISO 4-compliant
VWR® 16" sterile nitrile gloves are essential for cleanroom environments that require heightened protection, particularly in the pharmaceutical, biotech, and electronics industries. With their extended 16" length, these gloves offer enhanced forearm coverage, reducing contamination risks during high-sensitivity tasks. ISO Class 4-compliant and recommended for ISO Class 4 or less cleanroom environments, they provide a superior solution for maintaining stringent sterility and cleanliness, ensuring a safer barrier for users handling sensitive materials in contamination-controlled settings.
Packaging: Each bag contains 100 cleanroom gloves, ensuring optimal hygiene and contamination control. The gloves are packaged in a cleanroom environment, maintaining sterility and integrity. Each case includes 10 bags, totaling 1000 gloves per case. This packaging design allows for easy access and storage while ensuring that all gloves remain clean and uncontaminated until ready for use.