VWR® Cleanroom 16" Nitrile Non Sterile Gloves
Supplier: VWR International
76557-788 76557-790 76557-792 76557-794 76557-796 76557-798
76557-788CS 696.46 CAD
76557-788 76557-790 76557-792 76557-794 76557-796 76557-798
VWR® Cleanroom 16" Nitrile Non Sterile Gloves
Gloves
VWR® 16" non sterile nitrile gloves offer reliable, durable, and comfortable protection for cleanroom applications. Ideal for pharmaceutical, biotech, and electronics industries, these gloves meet the demands of controlled environments by providing excellent barrier protection and comfort in critical tasks.
  • Designed for elbow length protection
  • Ambidextrous, designed with micro textured fingers and palm, beaded cuff
  • Not made from natural rubber latex
  • Static-dissipative
  • Single-use only, ISO 4-compliant
VWR® 16" non sterile nitrile gloves are essential for cleanroom environments requiring extra protection, such as those in pharmaceutical, biotech, and electronics industries. Their extended 16" length provides enhanced forearm coverage, reducing contamination risks and ensuring a safer barrier during high-sensitivity tasks. ISO 4-compliant, these gloves are particularly beneficial for applications demanding cleanliness, offering an ideal solution for users handling sensitive materials in controlled environments that prioritize cleanliness and contamination control.
Packaging: Each bag contains 100 cleanroom gloves, ensuring optimal hygiene and contamination control. The gloves are packaged in a cleanroom environment, maintaining integrity. Each case includes 10 bags, totaling 1000 gloves per case. This packaging design allows for easy access and storage while ensuring that all gloves remain clean and uncontaminated until ready for use.