SmartDrop™ X Spectrophotometer
Supplier: Ward's
NS1010 NS1010-E
470346-300EA 19385.99 CAD
470346-300 470346-312
SmartDrop™ X Spectrophotometer
Spectrophotometers and Photometers Biophotometers
The SmartDrop™ series of nano spectrophotometers provides quick and easy quantification of small samples.
  • Full-spectrum absorbance scans from 200 to 800 nm
  • Quickly calculates and displays 260/280 and 260/230 ratios
  • Long-life Xenon lamp with precision monochromator
  • Auto-ranging path lengths of 0.2 and 1.0 mm
  • Detect up to 4500 ng/µl of dsDNA without dilution
These spectrophotometers utilize a small sample volume which eliminates the need for dilutions, providing more accurate results. Each unit includes a full color, 7"
touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computer is required.
SmartDrop™ L is the affordable choice for laboratories needing to determine nucleic acid and protein concentrations and purity of samples. The LED light source and silicon photocell sensor provide readings at 260 and 280 nm. Samples from 1.0 to 2.0 µl are loaded directly on the pedestal. Results are provided in about 5 seconds. The software automatically calculates and displays 260/280 ratios.
The resulting data can be printed, saved in the SmartDrop™'s onboard memory or exported via USB as an Excel file. All SmartDrop™ models include an integrated printer. For OD600 readings, each model includes a cuvette port and two optical glass cuvettes.