SmartDrop™ L Spectrophotometer
Supplier: Ward's
NS1000 NS1000-E
470346-306EA 10849.99 CAD
470346-306 470346-316
SmartDrop™ L Spectrophotometer
Spectrophotometers and Photometers Biophotometers
The SmartDrop™ series of nano spectrophotometers provides quick and easy quantification of small samples.
  • Ideal for basic DNA/RNA/protein quantification and purity
  • Affordable LED and silicon detector technology
  • Fixed 0.5 mm path length, detect up to 2500 ng/µl of dsDNA
These spectrophotometers utilize a small sample volume which eliminates the need for dilutions, providing more accurate results. Each unit includes a full color, 7"
touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computerMore Product Information