S-protein agarose, MilliporeSigma
Supplier: MilliporeSigma
69704-3 69704-4
CA80031-014EA 351.09 CAD
CA80031-014 CA80031-016
S-protein agarose, MilliporeSigma
Protein Purification Protein Purification Resins
S-protein agarose specifically retains STag™ fusion proteins. The capacity of S-protein agarose varies and is based on the size and folding characteristics of a given target protein.
Binding can be performed in a column or batch mode. Elution is possible using slightly denaturing conditions or using a specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins.