Ion exchange chromatography media, Ion Exchanger I
Supplier: MilliporeSigma
1.04765.5000 1.04765.5000 1.04765.0500 1.04765.0500
52428-634EA 1429.73 CAD
52428-634 52428-632 CA1.04765.5000 CA1.04765.0500
Ion exchange chromatography media, Ion Exchanger I
Protein Purification Protein Purification Resins
Ion Exchanger I is a strong acid cation exchangers widely used in separation of noble earths and amino acids, water treatment, analysis and food industry. The bond strength of equivalent ions increases with decreasing diameter of the hydrated ion.
Maximum working temperature: 120 °C
Exchange capacity: >1,7 mval/ml