Innovating Science® Coliform Test Powder Kits
Supplier: ALDON CORP SE
IS5055 IS5056
470343-534EA 20.49 CAD
470343-534 470343-536
Innovating Science® Coliform Test Powder Kits
Media for Microbiology Dehydrated Media for Microbiology
Easy way to test water for coliforms.
  • One water test for presence of coliforms
Simply add a small amount of collected water to the sample tube and incubate for 24 to 48 hours. A distinct color change from purple to yellow indicates that coliforms are present.