VWR® Salmon Cleanroom Paper
Supplier: VWR International
76547-894 76547-910 76547-912
76547-894CS 483.18 CAD
76547-894 76547-910 76547-912
VWR® Salmon Cleanroom Paper
Paper
Latex-Free paper has been injected with a synthetic copolymer to substantially reduce particle generation.
  • Exceptional thickness and strength, with excellent heat resistance
  • High opacity and 'class 10 certified'
Ideal for use in printers and high speed photocopies.
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Materials,Waste Reducing