PUREZERO* MARIN-XTRA* Nitrile Exam Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
LFS121LG LFS121MD LFS121SM LFS121XL LFS121XS
76554-012BX 32.15 CAD
76554-012 76554-032 76554-014 76554-024 76554-036
PUREZERO* MARIN-XTRA* Nitrile Exam Gloves, Halyard
Gloves
Designed for use in laboratories, research environments, and clean areas with applications in pharmaceutical, medical device manufacturing, biotechnology, and food processing/handling.
  • Features a SMARTPULL* Dispenser to control dispensing and lessen waste
  • Powder-free and natural rubber latex-free
  • Compliant for food handling applications
  • AQL 1.0 (exceeds the ASTM standard of AQL 2.5 for critical defects or pinholes)
  • Ambidextrous
  • Beaded cuffs to prevent roll-down
Lab workers may spend hours every day dealing with a variety of chemicals, some with the potential to impact their health and wellbeing. Therefore, workers should be wearing gloves that are highly protective and designed specifically for Life Science applications. To meet your these high standards, gloves have to be thoroughly tested, reliably supplied, and sustainably manufactured.
These gloves undergo comprehensive testing and standards, resulting in above industry average AQL (1.0) and 3 levels of color-coded protection with increasing thickness. These gloves are tested against 57 Chemicals, up to 14 Chemotherapy drugs, and Fentanyl. They are not made with natural rubber latex, for protection against Type IV allergies.
The Safeskin Glove Manufacturing facility (Thailand) uses 92% renewable energy. PUREZERO® Life Science Glove dispensers are made with up to 85% recycled materials, and cases are made with up to 99% recycled pulp. The innovative SMARTPULL* Dispenser1 reduces total waste by an average of 38,2% compared to a leading competitor. PUREZERO®* Life Science Gloves are manufactured in a facility that complies with the requirements of ISO 14001: 2015 - Environmental Management System.