Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
ADP-FLG001 FLO-FLG114.6 FLG.SP
76554-042EA 1666.9 CAD
76554-042 76638-932 77532-274
Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices, Oxford Nanopore Technologies
Flow Cells
Flongle is an adapter for MinION and GridION devices that enables direct, real-time DNA sequencing on smaller, single-use Flongle Flow Cells.
  • The most rapid, accessible, and cost-efficient nanopore sequencing system for smaller or more frequently performed assays and experiments
  • Each Flongle Flow can deliver 1–2 Gb of real-time sequencing data
The Flongle Adapter allows MinION and GridION devices to run smaller, single-use Flongle Flow Cells, typically generating 1–2 Gb of data per flow cell. Users can simply insert a Flongle Flow Cell into theMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.