Revolve™ Sustainable Pre-wetted Cleanroom Wipes, Texwipe®
Supplier: Texwipe
TX1704P TX1709P TX1712P
76472-840BG 57.4 CAD
76472-840 76472-842 76472-844
Revolve™ Sustainable Pre-wetted Cleanroom Wipes, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Wipe and clean surfaces, equipment and parts
  • Apply and remove lubricants, adhesives, residues and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol,
More Product Information