BPX90 Capillary Columns, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
054570 054580 054590 054596 054583
76409-334EA 769.37 CAD
76409-334 76409-336 76409-338 76409-340 76409-342
BPX90 Capillary Columns, Trajan Scientific and Medical
Chromatography Columns
Ideal for fast separation of fragrances, aromatics, petrochemical, pesticides, PCBs, and isomers of Fatty Acid Methyl Esters (FAMEs).
90% cyanopropyl polysilphenylene-siloxane.
Max. temperature: 80 to 280 °C.
Suitable replacement for: Unique to SGE.