University Microscopes with LCD Tablet, National Optical
Supplier: National Optical
BTI2-169-SP BTI2-169-ASC BTI2-169-P BTI1-169-SP
470337-682EA 3434.9 CAD
470337-682 470337-684 470337-686 470337-688
University Microscopes with LCD Tablet, National Optical
Microscopes
The Tablet series incorporates a wide range of features designed for university and laboratory use.
  • Connect 5 WiFi devices at one time
  • DIN standard 4×(0.10 N.A.), 10×(0.25 N.A.), 40×R (0.65 N.A.) and 100×R oil immersion (1.25 N.A.) plan objectives
  • All are parfocalled, parcentered and color-coded
  • 3W LED illumination with variable rheostat control
  • Large rackless 140×135mm (5½×5¼") stage; built-in, low profile, ball-bearing mounted mechanical
Combining the features of National's 169 advanced microscope with an Android based tablet system offers unlimitedMore Product Information