Masterflex® Union Fittings, Hose Barb, Tee, PVDF, Avantor®
Supplier: VWR International
MFLX40615-00 MFLX40615-01 MFLX40615-02 MFLX40615-03 MFLX40615-04 MFLX40615-05 MFLX40615-06 MFLX40615-07 MFLX40615-08 MFLX40615-09 MFLX40615-11 MFLX40615-12 MFLX40615-13 MFLX40615-14 MFLX40615-15 MFLX40615-16 MFLX40615-17 MFLX40615-18 MFLX40615-19 MFLX40615-20
MFLX40615-00EA 118.03 CAD
MFLX40615-00 MFLX40615-01 MFLX40615-02 MFLX40615-03 MFLX40615-04 MFLX40615-05 MFLX40615-06 MFLX40615-07 MFLX40615-08 MFLX40615-09 MFLX40615-11 MFLX40615-12 MFLX40615-13 MFLX40615-14 MFLX40615-15 MFLX40615-16 MFLX40615-17 MFLX40615-18 MFLX40615-19 MFLX40615-20
Masterflex® Union Fittings, Hose Barb, Tee, PVDF, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).