Masterflex® Adapter Fittings, Hose Barb to Threaded Adapter, Straight, PVDF, Avantor®
Supplier: VWR International
MFLX40614-07 MFLX40614-08 MFLX40614-09 MFLX40614-10 MFLX40614-11 MFLX40614-12 MFLX40614-13 MFLX40614-14 MFLX40614-16 MFLX40614-17 MFLX40614-18 MFLX40614-19 MFLX40614-20 MFLX40614-21 MFLX40614-22 MFLX40614-23 MFLX40614-24 MFLX40614-25 MFLX40614-26 MFLX40614-27 MFLX40614-28 MFLX40614-29 MFLX40614-30 MFLX40614-31 MFLX40614-32 MFLX40614-33 MFLX40614-34 MFLX40614-35 MFLX40614-15 MFLX30704-07
MFLX40614-07EA 40.29 CAD
MFLX40614-07 MFLX40614-08 MFLX40614-09 MFLX40614-10 MFLX40614-11 MFLX40614-12 MFLX40614-13 MFLX40614-14 MFLX40614-16 MFLX40614-17 MFLX40614-18 MFLX40614-19 MFLX40614-20 MFLX40614-21 MFLX40614-22 MFLX40614-23 MFLX40614-24 MFLX40614-25 MFLX40614-26 MFLX40614-27 MFLX40614-28 MFLX40614-29 MFLX40614-30 MFLX40614-31 MFLX40614-32 MFLX40614-33 MFLX40614-34 MFLX40614-35 MFLX40614-15 MFLX30704-07
Masterflex® Adapter Fittings, Hose Barb to Threaded Adapter, Straight, PVDF, Avantor®
Fittings and Connectors
Quick and easy installation with no tools required.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). PVDF material features excellent chemical resistance with a wide temperature range. Sterilize via ethylene oxide or autoclave.