Pulsafeeder Seven-Day Timer Pumps
Supplier: EQUIP SOLUTIONS
LC64BA-VTC1 LC13BA-VTC1 LC14BA-VTC1
MFLX74145-20EA 1343 CAD
MFLX74145-20 MFLX74145-00 MFLX74145-10
Pulsafeeder Seven-Day Timer Pumps
Pumps Diaphragm Pumps
Reliable metering performance—guided dual check valves provide precise seating and excellent priming and suction lift.
  • PVC and PTFE construction with external control
Seven-day timer pumps provide a simple and cost effective metering system for timed applications. Pumps offer eight on/eight off programs per week for maximum adjustability. These pumps feed chemical products on a timed schedule and are commonly used in biocide feed applications. Feed cycle is initiated and controlled by programmable timer.