Pulsafeeder Manual Control Metering Pumps
Supplier: EQUIP SOLUTIONS
LPG5SA-VTC3-XXX LD02SA-VTC1-XXX LD03SA-VTC1-XXX LD54SA-VTC1-XXX MFLX74142-20 MFLX74142-30 MFLX78118-20
MFLX74142-10EA 2512.2 CAD
MFLX74142-10 MFLX78118-00 MFLX78118-10 MFLX78118-30 MFLX74142-20 MFLX74142-30 MFLX78118-20
Pulsafeeder Manual Control Metering Pumps
Pumps Diaphragm Pumps
Reliable metering performance – guided dual check valves provide precise seating and excellent priming and suction lift.
  • PVC and PTFE construction with external control
  • Ideal for water treatment and chemical feed applications
Manual control pumps feature adjustable stroke rate and length, reliable timing circuits, and circuit protection from voltage and current upsets. Leak-free bleed valves provide safe and easy priming. Pumps are water resistant and can be installed outdoors.
Delivery information: Includes injection/back pressure valve assembly,More Product Information