Eldex High-Pressure Piston Pumps
Supplier: Eldex Corporation
1018 1005 1384
MFLX07143-86EA 5269.3 CAD
MFLX07143-86 MFLX07143-90 MFLX07143-70
Eldex High-Pressure Piston Pumps
Pumps Piston Pumps
Feature micrometer flow adjustment—deliver exact quantities of liquid with minimal pulsation.
  • Withstand up to 5000 psi outlet pressure
  • Dual-piston models have independent piston control—pump a single liquid, or pump two different liquids at a proportional rate
  • Brushless synchronous motors
Inlet accepts 1/8" OD tubing; outlet accepts 1/16" OD high-pressure SS tubing (order separately). Decrease pulsation with pulse dampener model 07115-55.
What's included: All models come with a sturdy aluminum housing and a 7-ft, three-wire cord (115 VAC models have a plug; 220 VAC models have a European plug).
FLUOROLOY G® The Fluorocarbon Co.
KEL-F® 3M Co.