Hand-Operated Chemical-Resistant Drum Pumps
Supplier: BURKLE INC
5606-0600 5606-1000 5606-1001 MFLX06043-97
MFLX06043-96EA 1301.92 CAD
MFLX06043-96 MFLX06043-98 MFLX06043-99 MFLX06043-97
Hand-Operated Chemical-Resistant Drum Pumps
Pumps Drum/Transfer Pumps
Safely transfer aggressive chemicals.
  • PTFE and FEP wetted materials withstand harsh chemicals
  • Stopcock contains hazardous vapors and prevents dripping
These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).