Remote-Control Diaphragm Liquid Dosing Pumps
Supplier: KNF Neuberger Inc.
167533/160160 MFLX78167-00 MFLX78167-01 MFLX78167-02 MFLX78167-05 MFLX78167-07 MFLX78167-10
MFLX78167-09EA 4392.4 CAD
MFLX78167-09 MFLX78167-00 MFLX78167-01 MFLX78167-02 MFLX78167-05 MFLX78167-07 MFLX78167-10
Remote-Control Diaphragm Liquid Dosing Pumps
Pumps Diaphragm Pumps
Uniformity of discharge ensures low-pulsation transfer process.
  • Quiet, reliable, accurate, and virtually maintenance-free
  • Pumps can run dry continuously without damage
  • Versatile pumps handle flow rates up to 100 ml/min and volumes up to 1000 mL
Simple and intuitive function makes the liquid dosing pump an accurate and reliable solution for your laboratory dosing applications. User-friendly design provides a quiet, dry-running, and self-priming dosing operation. Easy calibration and cycle mode allow for uninterrupted and virtually maintenance-free dosing.
This pump features a PTFE-covered diaphragm and IP65-rated splash protection, making it resistant to chemical attack and adaptable to fluid characteristics.
Unique drive system ensures maximum uniformity of discharge resulting in a low-pulsation transfer process. Pumps feature either local or external control capabilities including start/stop and reset/prime via TTL contact as well as output signal fault alarm. Models available with a RS-232 port for connectivity.
Delivery information: Includes 5-ft (1.5-m) power cord and two 3-ft (1-m) pieces of FEP inlet/outlet tubing (1/8" OD x 1/16" ID) with UNF 1/4"-28 fittings. Models with RS-232 port also include a cable with D-Sub-9 connector.