Pulsafeeder Economical Peristaltic Pumps
Supplier: EQUIP SOLUTIONS
XP015LAHX XP030LALX MFLX74130-02 MFLX74130-11 MFLX74130-12 MFLX74130-13 MFLX74130-18 MFLX74130-24 MFLX74130-26
MFLX74130-04EA 1415.68 CAD
MFLX74130-04 MFLX74130-07 MFLX74130-02 MFLX74130-11 MFLX74130-12 MFLX74130-13 MFLX74130-18 MFLX74130-24 MFLX74130-26
Pulsafeeder Economical Peristaltic Pumps
Pumps Peristaltic Pumps
Easy maintenance and no contamination—tubing is the only wetted part.
  • Easy tube change-out minimizes downtime between tubing set changes
  • Inherent degassing function and rugged gear train offer many years of reliable service
These simple-to-use, self-priming peristaltic pumps deliver worry-free dosing.
Delivery information: Includes pump head, two tubing assemblies, and a 6-ft (1.8-m) power cord.