Foot-Operated Pump Dispensers for Barrels, Canisters, and Tanks
Supplier: BURKLE INC
5000-1800 5000-2200 5000-4016 MFLX06040-31 MFLX06040-43
MFLX06040-33EA 243.32 CAD
MFLX06040-33 MFLX06040-35 MFLX06040-39 MFLX06040-31 MFLX06040-43
Foot-Operated Pump Dispensers for Barrels, Canisters, and Tanks
Pumps Drum/Transfer Pumps
Keep hands-free for safe dispensing.
  • Large-volume foot-operated bellows for high pumping capacity
  • Universal bung constructed of soft PVC for 1-3/16" to 2-3/4" (30 to 70 mm) openings
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
Safely transfer liquids keeping both hands on the container you are filling. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.