LinkTech 20CB Series Quick-Disconnect Fitting Body, BSPT(M) without Valve
Supplier: VWR International
20CB-S1-02BSPT
MFLX41404-40EA 162.74 CAD
MFLX41404-40
LinkTech 20CB Series Quick-Disconnect Fitting Body, BSPT(M) without Valve
Fittings and Connectors
Chrome-plated brass construction withstands higher operating pressures.
  • Audible click lets you know the coupling is secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
These food-grade chrome-plated brass fittings are well suited for food and beverage applications. The 1/8” flow size bodies and inserts are durable and can withstand operating pressures up to 250 psi (17.2 bar). Intuitive thumb-latch design makes disconnecting fittings quick and easy with one hand.
You can easily mix and match valved and straight-through fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.