LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Body, NPT(M) with Valve
Supplier: VWR International
20ACV-SB1-04
MFLX16360-06EA 254.38 CAD
MFLX16360-06
LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Body, NPT(M) with Valve
Fittings and Connectors
Simply press the stainless steel thumb latch to disconnect the fittings.
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.