Cylindrical Barbed Cross Connectors, PVDF
Supplier: BURKLE INC
8717-0004 8717-0006 8717-0010 8717-0012 8717-0014 MFLX06044-31
MFLX06044-28EA 29.7 CAD
MFLX06044-28 MFLX06044-29 MFLX06044-32 MFLX06044-33 MFLX06044-34 MFLX06044-31
Cylindrical Barbed Cross Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed cross connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.