Bürkle Cylindrical Barbed Y-Connectors, PVDF
Supplier: BURKLE INC
8712-0004 8712-0006 8712-0008 8712-0010 8712-0012 MFLX06044-21
MFLX06044-15EA 27.97 CAD
MFLX06044-15 MFLX06044-16 MFLX06044-17 MFLX06044-18 MFLX06044-19 MFLX06044-21
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed Y connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.