Bürkle Cylindrical Barbed Tee Connectors, PVDF
Supplier: BURKLE INC
8711-0004 8711-0006 8711-0008 8711-0010 MFLX06044-14 MFLX06044-13
MFLX06044-08EA 19.28 CAD
MFLX06044-08 MFLX06044-09 MFLX06044-11 MFLX06044-12 MFLX06044-14 MFLX06044-13
Bürkle Cylindrical Barbed Tee Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed tee connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.