Cylindrical Barbed Straight Connectors, PVDF
Supplier: BURKLE INC
8710-0006 8710-0010 8710-0014 8710-0004 8710-0008 8710-0012
MFLX06044-01EA 13.75 CAD
MFLX06044-01 MFLX06044-03 MFLX06044-02 MFLX06044-05 MFLX06044-04 MFLX06044-06
Cylindrical Barbed Straight Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed straight connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.