Value Plastics Sanitary Adapters, Barb to Sanitary Clamp, PVDF
Supplier: Avantor Fluid Handling
SFMX670-J1A SFMN655-J1A MFLX31513-63 MFLX31513-69 MFLX40617-52BLK MFLX40617-53BLK SFMN680-J2A (100PK) MFLX40617-57BLK MFLX40617-59BLK MFLX40617-60BLK MFLX40617-51BLK MFLX40617-56BLK SFMXC-J2A (50PK) Y680-6005 (50PK) MFLX40617-87BLK
MFLX31513-66EA 77.89 CAD
MFLX31513-66 MFLX31513-67 MFLX31513-61 MFLX31513-62 MFLX31513-65 MFLX31513-63 MFLX31513-69 MFLX40617-52BLK MFLX40617-53BLK MFLX40617-54BLK MFLX40617-57BLK MFLX40617-59BLK MFLX40617-60BLK MFLX40617-51BLK MFLX40617-56BLK MFLX40617-61BLK MFLX40617-80BLK MFLX40617-87BLK
Value Plastics Sanitary Adapters, Barb to Sanitary Clamp, PVDF
Fittings and Connectors
Create a secure connection in your sanitary fitting system.
  • Constructed of durable PVDF
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow without particulate build-up. There are no sharp edges in the flow path to make cleaning very easy. These adapters offer good chemical resistance and may be sterilized by gamma irradiation, EtOH, or autoclave. Material meets the requirements for USP Class VI, ADCF, FDA, and are BPA-free.